2'-Chloro-1-isopropyl-1,2,3,4,5,6-hexahydro[4,4']bipyridinyl

Supplier: Matrix Scientific

067386-250MG
101933-890EA 558.46 USD
101933-890
2'-Chloro-1-isopropyl-1,2,3,4,5,6-hexahydro[4,4']bipyridinyl
2'-Chloro-1-isopropyl-1,2,3,4,5,6-hexahydro[4,4']bipyridinyl

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂
MDL Number: MFCD19691492
CAS Number: 1316225-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR