2-Bromo-6-(1-isopropylpyrrolidin-3-ylmethyl)pyridine

Supplier: Matrix Scientific

067384-250MG
101933-886EA 638.46 USD
101933-886
2-Bromo-6-(1-isopropylpyrrolidin-3-ylmethyl)pyridine
2-Bromo-6-(1-isopropylpyrrolidin-3-ylmethyl)pyridine

Hazard: Irritant

Formula: C₁₃H₁₉BrN₂
MW: 283.22 g/mol
MDL Number: MFCD19691622
CAS Number: 1316222-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR