2-Bromo-6-(1-isopropylpyrrolidin-2-yl)pyridine

Supplier: Matrix Scientific

067382-250MG
101933-882EA 558.46 USD
101933-882
2-Bromo-6-(1-isopropylpyrrolidin-2-yl)pyridine
2-Bromo-6-(1-isopropylpyrrolidin-2-yl)pyridine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂
MDL Number: MFCD19691489
CAS Number: 1316225-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR