1-[3-(3-Chloropyrazin-2-yl)piperidin-1-yl]ethanone
Supplier: Matrix Scientific
067381-250MG
101933-880EA
558.46
USD
101933-880
1-[3-(3-Chloropyrazin-2-yl)piperidin-1-yl]ethanone
1-[3-(3-Chloropyrazin-2-yl)piperidin-1-yl]ethanone
Hazard: Irritant
Formula:
C₁₁H₁₄ClN₃O |
MDL Number:
MFCD19691518 CAS Number: 1316220-01-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...