1-[2-(5-Bromo-4-methylthiazol-2-yl)pyrrolidin-1-yl]ethanone

Supplier: Matrix Scientific

067379-250MG
101933-876EA 476.92 USD
101933-876
1-[2-(5-Bromo-4-methylthiazol-2-yl)pyrrolidin-1-yl]ethanone
1-[2-(5-Bromo-4-methylthiazol-2-yl)pyrrolidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₀H₁₃BrN₂OS
MDL Number: MFCD16621972
CAS Number: 1361112-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR