2-Chloro-4-[2-(1-methanesulfonylpiperidin-2-yl)ethyl]pyridine

Supplier: Matrix Scientific

067376-250MG
101933-870EA 558.46 USD
101933-870
2-Chloro-4-[2-(1-methanesulfonylpiperidin-2-yl)ethyl]pyridine
2-Chloro-4-[2-(1-methanesulfonylpiperidin-2-yl)ethyl]pyridine

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂S
MDL Number: MFCD19691548
CAS Number: 1316220-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR