2-Chloro-4-[2-(1-methanesulfonylpyrrolidin-2-yl)ethyl]pyridine

Supplier: Matrix Scientific

067375-250MG
101933-868EA 638.46 USD
101933-868
2-Chloro-4-[2-(1-methanesulfonylpyrrolidin-2-yl)ethyl]pyridine
2-Chloro-4-[2-(1-methanesulfonylpyrrolidin-2-yl)ethyl]pyridine

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂S
MDL Number: MFCD19691635
CAS Number: 1316222-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR