1-[4-(6-Chloropyrazin-2-ylmethyl)azepan-1-yl]ethanone

Supplier: Matrix Scientific

067371-250MG
101933-860EA 638.46 USD
101933-860
1-[4-(6-Chloropyrazin-2-ylmethyl)azepan-1-yl]ethanone
1-[4-(6-Chloropyrazin-2-ylmethyl)azepan-1-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O
MDL Number: MFCD19691629
CAS Number: 1316221-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR