1-[4-(3-Chloropyrazin-2-ylmethyl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067365-250MG
101933-848EA 558.46 USD
101933-848
1-[4-(3-Chloropyrazin-2-ylmethyl)piperidin-1-yl]ethanone
1-[4-(3-Chloropyrazin-2-ylmethyl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₂H₁₆ClN₃O
MDL Number: MFCD19691521
CAS Number: 1316227-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR