1-[4-(6-Chloro-2-methylpyrimidin-4-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one

Supplier: Matrix Scientific

067363-250MG
101933-844EA 558.46 USD
101933-844
1-[4-(6-Chloro-2-methylpyrimidin-4-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one
1-[4-(6-Chloro-2-methylpyrimidin-4-yl)piperidin-1-yl]-2,2-dimethylpropan-1-one

Hazard: Irritant

Formula: C₁₅H₂₂ClN₃O
MDL Number: MFCD19691587
CAS Number: 1316217-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR