1-[4-(6-Chloropyridazin-3-ylmethyl)azepan-1-yl]ethanone

Supplier: Matrix Scientific

067362-250MG
101933-842EA 638.46 USD
101933-842
1-[4-(6-Chloropyridazin-3-ylmethyl)azepan-1-yl]ethanone
1-[4-(6-Chloropyridazin-3-ylmethyl)azepan-1-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O
MDL Number: MFCD19691631
CAS Number: 1316227-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR