1-[4-(5-Bromopyrazin-2-ylmethyl)azepan-1-yl]ethanone

Supplier: Matrix Scientific

067357-250MG
101933-832EA 558.46 USD
101933-832
1-[4-(5-Bromopyrazin-2-ylmethyl)azepan-1-yl]ethanone
1-[4-(5-Bromopyrazin-2-ylmethyl)azepan-1-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₈BrN₃O
MDL Number: MFCD19691528
CAS Number: 1316217-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR