1-[4-(5-Bromopyrazin-2-ylmethyl)piperidin-1-yl]ethanone

Supplier: Matrix Scientific

067355-250MG
101933-828EA 638.46 USD
101933-828
1-[4-(5-Bromopyrazin-2-ylmethyl)piperidin-1-yl]ethanone
1-[4-(5-Bromopyrazin-2-ylmethyl)piperidin-1-yl]ethanone

Hazard: Irritant

Formula: C₁₂H₁₆BrN₃O
MDL Number: MFCD19691527
CAS Number: 1316220-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR