1-[4-(4-Chloro-6-methylpyrimidin-2-yl)piperidin-1-yl]-2-methoxyethanone

Supplier: Matrix Scientific

067354-250MG
101933-826EA 558.46 USD
101933-826
1-[4-(4-Chloro-6-methylpyrimidin-2-yl)piperidin-1-yl]-2-methoxyethanone
1-[4-(4-Chloro-6-methylpyrimidin-2-yl)piperidin-1-yl]-2-methoxyethanone

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O₂
MDL Number: MFCD19691589
CAS Number: 1316220-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR