Azetidin-3-ylpiperidin-1-ylmethanonetrifluoroacetic acid salt
Supplier: Matrix Scientific
067325-250MG
101933-768EA
476.92
USD
101933-768
Azetidin-3-ylpiperidin-1-ylmethanonetrifluoroacetic acid salt
Azetidin-3-ylpiperidin-1-ylmethanonetrifluoroacetic acid salt
Hazard: Irritant
Formula:
C₁₁H₁₇F₃N₂O₃ MW: 282.26 g/mol |
MDL Number:
MFCD21605778 CAS Number: 1361114-44-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...