3-Pyrazol-1-ylmethylpiperidine

Supplier: Matrix Scientific

067312-250MG
101933-742EA 476.92 USD
101933-742
3-Pyrazol-1-ylmethylpiperidine
3-Pyrazol-1-ylmethylpiperidine

Hazard: Irritant

Formula: C₉H₁₅N₃
MDL Number: MFCD15202327
CAS Number: 1211450-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR