2-(5-Bromopyridin-2-yl)-4-isopropylmorpholine

Supplier: Matrix Scientific

067295-250MG
101933-708EA 638.46 USD
101933-708
2-(5-Bromopyridin-2-yl)-4-isopropylmorpholine
2-(5-Bromopyridin-2-yl)-4-isopropylmorpholine

Hazard: Irritant

Formula: C₁₂H₁₇BrN₂O
MDL Number: MFCD19691605
CAS Number: 1316227-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR