(3aS,8aS)-4-(2-Trifluoromethylbenzyl)octahydropyrrolo[3,2-b]azepin-5-one

Supplier: Matrix Scientific

067294-250MG
101933-706EA 687.69 USD
101933-706
(3aS,8aS)-4-(2-Trifluoromethylbenzyl)octahydropyrrolo[3,2-b]azepin-5-one
(3aS,8aS)-4-(2-Trifluoromethylbenzyl)octahydropyrrolo[3,2-b]azepin-5-one

Hazard: Irritant

Formula: C₁₆H₁₉F₃N₂O
MDL Number: MFCD19691660
CAS Number: 1442080-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR