Morpholine-2-carboxylic acid[2-(3,5-dimethyl-1H-pyrazol-4-yl)ethyl]amidedihydrochloride

Supplier: Matrix Scientific

067293-500MG
101933-704EA 623.08 USD
101933-704
Morpholine-2-carboxylic acid[2-(3,5-dimethyl-1H-pyrazol-4-yl)ethyl]amidedihydrochloride
Morpholine-2-carboxylic acid[2-(3,5-dimethyl-1H-pyrazol-4-yl)ethyl]amidedihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂Cl₂N₄O₂
MW: 325.24 g/mol
MDL Number: MFCD21605769
CAS Number: 1361111-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR