Morpholine-2-carboxylic acid(1-ethyl-3,5-dimethyl-1H-pyrazol-4-yl)amide dihydrochloride

Supplier: Matrix Scientific

067292-500MG
101933-702EA 623.08 USD
101933-702
Morpholine-2-carboxylic acid(1-ethyl-3,5-dimethyl-1H-pyrazol-4-yl)amide dihydrochloride
Morpholine-2-carboxylic acid(1-ethyl-3,5-dimethyl-1H-pyrazol-4-yl)amide dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂Cl₂N₄O₂
MW: 325.24 g/mol
MDL Number: MFCD21605768
CAS Number: 1361113-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR