3-Biphenyl-2-ylmethyl-1-propylpiperazin-2-one

Supplier: Matrix Scientific

067289-250MG
101933-696EA 558.46 USD
101933-696
3-Biphenyl-2-ylmethyl-1-propylpiperazin-2-one
3-Biphenyl-2-ylmethyl-1-propylpiperazin-2-one

Hazard: Irritant

Formula: C₂₀H₂₄N₂O
MDL Number: MFCD21605765
CAS Number: 1361111-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR