3-Biphenyl-2-ylmethyl-1-ethylpiperazin-2-one

Supplier: Matrix Scientific

067288-250MG
101933-694EA 558.46 USD
101933-694
3-Biphenyl-2-ylmethyl-1-ethylpiperazin-2-one
3-Biphenyl-2-ylmethyl-1-ethylpiperazin-2-one

Hazard: Irritant

Formula: C₁₉H₂₂N₂O
MDL Number: MFCD21605764
CAS Number: 1361111-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR