2-(S)-1-Pyrrolidin-2-ylmethyl-2,5,6,7,8,9-hexahydro-cyclohepta[c]pyridazin-3-one

Supplier: Matrix Scientific

067282-250MG
101933-682EA 476.92 USD
101933-682
2-(S)-1-Pyrrolidin-2-ylmethyl-2,5,6,7,8,9-hexahydro-cyclohepta[c]pyridazin-3-one
2-(S)-1-Pyrrolidin-2-ylmethyl-2,5,6,7,8,9-hexahydro-cyclohepta[c]pyridazin-3-one

Hazard: Irritant

Formula: C₁₄H₂₁N₃O
MDL Number: MFCD16621960
CAS Number: 1361120-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR