(S)-2-Boc-aminomethylpyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-tert-butyl pyrrolidin-2-ylmethylcarbamate(S)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate

067240-1G
101933-622EA 104.62 USD
101933-622
(S)-2-Boc-aminomethylpyrrolidine ≥95%
(S)-2-Boc-aminomethylpyrrolidine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.2780 g/mol
MDL Number: MFCD06796572
CAS Number: 141774-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR