5-Cyano-2-methylpyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
6-Methylnicotinonitrile
, 5-Cyano-2-picoline
067239-5G
067239-1G
101933-620EA
89.23
USD
101933-620
101933-618
5-Cyano-2-methylpyridine ≥98%
5-Cyano-2-methylpyridine
Hazard: Irritant
Formula:
C₇H₆N₂ MW: 118.14 g/mol |
MDL Number:
MFCD00038042 CAS Number: 3222-48-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...