7-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥97%

Supplier: Matrix Scientific

067238-500MG
101933-616EA 310.77 USD
101933-616
7-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride ≥97%
7-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₄ClN
MDL Number: MFCD09026791
CAS Number: 41565-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR