2-Amino-6-chloro-3-pyridinecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-6-chloronicotinamide

067231-1G 067231-5G
101933-594EA 249.23 USD
101933-594 102551-698
2-Amino-6-chloro-3-pyridinecarboxamide ≥95%
2-Amino-6-chloro-3-pyridinecarboxamide

Hazard: Irritant

Formula: C₆H₆ClN₃O
MW: 171.59 g/mol
MDL Number: MFCD12024435
CAS Number: 64321-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR