(-)-Cotinine ≥97%

Supplier: Matrix Scientific

Synonyms: (S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one

067229-1G
101933-590EA 138 USD
101933-590
(-)-Cotinine ≥97%
(-)-Cotinine

Hazard: Irritant

Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Boiling Pt: 145…147 °C (3 mmHg)
MDL Number: MFCD00077696
CAS Number: 486-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR