2,2-Difluoro-1-phenylcyclopropanecarboxylic acid ≥97%

067228-500MG
101933-588EA 225.21 USD
101933-588
2,2-Difluoro-1-phenylcyclopropanecarboxylic acid ≥97%
2,2-Difluoro-1-phenylcyclopropanecarboxylic acid

Hazard: Irritant-Harmful/Lachrymator

Formula: C₁₀H₈F₂O₂
MW: 198.17 g/mol
Melting Pt: 100…103 °C
MDL Number: MFCD18434522
CAS Number: 156021-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR