Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate
Supplier: Matrix Scientific
Synonyms:
(-)-1-Ethyl-L-proline Ethyl Ester
, (S)-(-)-1-Ethyl-2-pyrrolidinecarboxylic Acid Ethyl Ester
, Et-Pro-OEt
064031-500MG
101926-854EA
364.62
USD
101926-854
Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate
Ethyl (S)-(-)-1-ethyl-2-pyrrolidinecarboxylate
CAS: 938-54-5
MDL Number: MFCD00191434
Molecular Formula: C9H17NO2
Molecular Weight: 171,24
Hazard: Irritant
Formula:
C₉H₁₇NO₂ MW: 171.24 g/mol |
MDL Number:
MFCD00191434 CAS Number: 938-54-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...