2-[(5-Methyl-1,3-benzoxazol-2-yl)methyl]-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

064024-500MG
101926-840EA 243.08 USD
101926-840
2-[(5-Methyl-1,3-benzoxazol-2-yl)methyl]-1H-isoindole-1,3(2H)-dione
2-[(5-Methyl-1,3-benzoxazol-2-yl)methyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₇H₁₂N₂O₃
MDL Number: MFCD16618481
CAS Number: 1228553-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR