2-tert-Butyl-1H-imidazole
Supplier: Matrix Scientific
Synonyms:
2-(tert-Butyl)-1H-imidazole
064023-500MG
101926-838EA
110.77
USD
101926-838
2-tert-Butyl-1H-imidazole
2-tert-Butyl-1H-imidazole
Hazard: Irritant
Formula:
C₇H₁₂N₂ MW: 124.19 g/mol |
MDL Number:
MFCD16618480 CAS Number: 36947-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...