1-Propylimidazole

Supplier: Matrix Scientific

Synonyms: 1-Propyl-1H-imidazole

064021-500MG
101926-834EA 364.62 USD
101926-834
1-Propylimidazole
1-Propylimidazole

Hazard: Irritant

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
MDL Number: MFCD00060494
CAS Number: 35203-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR