(1E,3E)-(Phenylhydrazono)malonaldehyde dioxime

Supplier: Matrix Scientific

064010-500MG
101926-814EA 243.08 USD
101926-814
(1E,3E)-(Phenylhydrazono)malonaldehyde dioxime
(1E,3E)-(Phenylhydrazono)malonaldehyde dioxime

Hazard: Irritant

Formula: C₉H₁₀N₄O₂
MDL Number: MFCD16618472
CAS Number: 41886-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR