(1-Isopropyl-1H-pyrazol-4-yl)methanol

Supplier: Matrix Scientific

063998-500MG
101926-792EA 364.62 USD
101926-792
(1-Isopropyl-1H-pyrazol-4-yl)methanol
(1-Isopropyl-1H-pyrazol-4-yl)methanol

Hazard: Irritant

Formula: C₇H₁₂N₂O
MDL Number: MFCD08059963
CAS Number: 1007542-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR