5-Fluoro-4-hydrazinyl-2-methoxypyrimidine

Supplier: Matrix Scientific

063996-500MG
101926-788EA 606.15 USD
101926-788
5-Fluoro-4-hydrazinyl-2-methoxypyrimidine
5-Fluoro-4-hydrazinyl-2-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₇FN₄O
CAS Number: 166524-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR