[2-Nitro-5-(propylthio)phenyl]amine

Supplier: Matrix Scientific

063985-500MG
101926-768EA 364.62 USD
101926-768
[2-Nitro-5-(propylthio)phenyl]amine
[2-Nitro-5-(propylthio)phenyl]amine

Hazard: Irritant

Formula: C₉H₁₂N₂O₂S
MW: 212.27 g/mol
MDL Number: MFCD00192355
CAS Number: 57780-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR