2-(4,5-Dihydro-1H-pyrazol-1-yl)ethanol

Supplier: Matrix Scientific

063978-500MG
101926-752EA 484.62 USD
101926-752
2-(4,5-Dihydro-1H-pyrazol-1-yl)ethanol
2-(4,5-Dihydro-1H-pyrazol-1-yl)ethanol

Hazard: Irritant

Formula: C₅H₁₀N₂O
MDL Number: MFCD16556126
CAS Number: 5677-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR