N-(1-Methylprop-2-yn-1-yl)cyclohexanamine

Supplier: Matrix Scientific

063971-500MG
101926-738EA 484.62 USD
101926-738
N-(1-Methylprop-2-yn-1-yl)cyclohexanamine
N-(1-Methylprop-2-yn-1-yl)cyclohexanamine

Hazard: Irritant

Formula: C₁₀H₁₇N
MDL Number: MFCD16618461
CAS Number: 86024-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR