N-1-Naphthyl-3-oxo-3-phenylpropanamide

Supplier: Matrix Scientific

063970-500MG
101926-736EA 243.08 USD
101926-736
N-1-Naphthyl-3-oxo-3-phenylpropanamide
N-1-Naphthyl-3-oxo-3-phenylpropanamide

Hazard: Irritant

Formula: C₁₉H₁₅NO₂
MDL Number: MFCD11974641
CAS Number: 20653-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR