N-2-Naphthyl-3-oxo-3-phenylpropanamide

Supplier: Matrix Scientific

063969-500MG
101926-734EA 243.08 USD
101926-734
N-2-Naphthyl-3-oxo-3-phenylpropanamide
N-2-Naphthyl-3-oxo-3-phenylpropanamide

Hazard: Irritant

Formula: C₁₉H₁₅NO₂
MDL Number: MFCD00712044
CAS Number: 17738-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR