4-(Dimethylamino)-3,5-dinitrobenzoic acid

Supplier: Matrix Scientific

063967-500MG
101926-730EA 364.62 USD
101926-730
4-(Dimethylamino)-3,5-dinitrobenzoic acid
4-(Dimethylamino)-3,5-dinitrobenzoic acid

Hazard: Irritant

Formula: C₉H₉N₃O₆
MDL Number: MFCD00024508
CAS Number: 82366-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR