3,3'-Diethoxybiphenyl-4,4'-diamine

Supplier: Matrix Scientific

063966-500MG
101926-728EA 243.08 USD
101926-728
3,3'-Diethoxybiphenyl-4,4'-diamine
3,3'-Diethoxybiphenyl-4,4'-diamine

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₂
MDL Number: MFCD05999095
CAS Number: 6264-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR