(3S,4S,5S,6R)-2-(Hydroxymethyl)-6-(1H-indol-1-yl)tetrahydro-2H-pyran-3,4,5-triol

Supplier: Matrix Scientific

063947-500MG
101926-688EA 243.08 USD
101926-688
(3S,4S,5S,6R)-2-(Hydroxymethyl)-6-(1H-indol-1-yl)tetrahydro-2H-pyran-3,4,5-triol
(3S,4S,5S,6R)-2-(Hydroxymethyl)-6-(1H-indol-1-yl)tetrahydro-2H-pyran-3,4,5-triol

Hazard: Irritant

Formula: C₁₄H₁₇NO₅
Melting Pt: 30 °C
MDL Number: MFCD16618451
CAS Number: 5059-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR