4-Methylisatin

Supplier: Matrix Scientific

Synonyms: 4-Methyl-1H-indole-2,3-dione

063945-500MG
101926-684EA 606.15 USD
101926-684
4-Methylisatin
4-Methylisatin

Hazard: Irritant

Formula: C₉H₇NO₂
MDL Number: MFCD00463404
CAS Number: 1128-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR