Ethyl-6-methoxy-1H-indole-2-carboxylate

Supplier: Matrix Scientific

063943-500MG
101926-680EA 400 USD
101926-680
Ethyl-6-methoxy-1H-indole-2-carboxylate
Ethyl-6-methoxy-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
MDL Number: MFCD01544121
CAS Number: 15050-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR