1-Acetyl-6-nitroindoline

Supplier: Matrix Scientific

063941-500MG
101926-676EA 146.15 USD
101926-676
1-Acetyl-6-nitroindoline
1-Acetyl-6-nitroindoline

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃
MDL Number: MFCD00022910
CAS Number: 22949-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR