[2-(1H-Pyrazol-1-ylmethyl)benzyl]amine

Supplier: Matrix Scientific

063937-500MG
101926-668EA 364.62 USD
101926-668
[2-(1H-Pyrazol-1-ylmethyl)benzyl]amine
[2-(1H-Pyrazol-1-ylmethyl)benzyl]amine

Hazard: Irritant

Formula: C₁₁H₁₃N₃
MDL Number: MFCD08245275
CAS Number: 878466-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR