Quinolin-8-ylmethanol

Supplier: Matrix Scientific

063935-500MG
101926-664EA 484.62 USD
101926-664
Quinolin-8-ylmethanol
Quinolin-8-ylmethanol

Hazard: Irritant

Formula: C₁₀H₉NO
MDL Number: MFCD00769488
CAS Number: 16032-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR