5-Phenyl-1H-1,2,4-triazole-3-carboxylic acid

Supplier: Matrix Scientific

063930-500MG
101926-654EA 606.15 USD
101926-654
5-Phenyl-1H-1,2,4-triazole-3-carboxylic acid
5-Phenyl-1H-1,2,4-triazole-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₇N₃O₂
MDL Number: MFCD16618449
CAS Number: 7200-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR